Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166313.1 | NP_001159785.1 | 7 - 8 | 873 | 97 | |
XM_005661544.3 | XP_005661601.1 | 1102 | 97 | |||
XM_013994971.2 | XP_013850425.1 | 1435 | 97 | |||
XM_013994972.2 | XP_013850426.1 | 1435 | 97 | |||
XM_021077311.1 | XP_020932970.1 | 1435 | 97 | |||
GenBank mRNA | AB499892.1 | - | 7 - 8 | 873 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |