Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167643.1 | NP_001161115.1 | 3 - 4 | 409 | 77 | |
GenBank mRNA | AB529450.1 | - | 3 - 4 | 409 | 77 | |
AK235587.1 | - | 3 - 4 | 673 | 77 | ||
AK395698.1 | - | 4 - 5 | 526 | 77 | ||
AY609406.1 | - | 3 - 4 | 639 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |