Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185138.1 | NP_001172067.1 | 1 - 2 | 164 | 92 | |
XM_005665780.3 | XP_005665837.1 | 448 | 92 | |||
XM_021098057.1 | XP_020953716.1 | 458 | 92 | |||
GenBank mRNA | AB154366.1 | - | 1 - 2 | 164 | 92 | |
AB294210.1 | - | 1 - 2 | 164 | 92 | ||
AK233944.1 | - | 1 - 2 | 167 | 92 | ||
AK234043.1 | - | 1 - 2 | 166 | 92 | ||
AK345069.1 | - | 1 - 2 | 167 | 92 | ||
AK345209.1 | - | 1 - 2 | 166 | 92 | ||
AK346577.1 | - | 1 - 2 | 166 | 92 | ||
AK346655.1 | - | 1 - 2 | 166 | 92 | ||
AK346802.1 | - | 1 - 2 | 166 | 92 | ||
AK346819.1 | - | 1 - 2 | 168 | 92 | ||
AK346962.1 | - | 1 - 2 | 166 | 92 | ||
AK346972.1 | - | 1 - 2 | 166 | 92 | ||
AK347038.1 | - | 1 - 2 | 169 | 92 | ||
AK347059.1 | - | 1 - 2 | 171 | 92 | ||
EU282350.1 | - | 1 - 2 | 168 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |