Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185139.1 | NP_001172068.1 | 3 - 4 | 448 | 64 | |
GenBank mRNA | AB529862.1 | - | 3 - 4 | 448 | 64 | |
AK240367.1 | - | 3 - 4 | 559 | 64 | ||
AK347608.1 | - | 3 - 4 | 523 | 64 | ||
AY609633.1 | - | 3 - 4 | 497 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |