Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM204A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185171.1 | NP_001172100.1 | 6 - 7 | 697 | 75 | |
XM_005671491.3 | XP_005671548.1 | 533 | 75 | |||
XM_005671493.3 | XP_005671550.1 | 449 | 75 | |||
XM_005671494.3 | XP_005671551.1 | 478 | 75 | |||
XM_005671498.3 | XP_005671555.1 | 411 | 75 | |||
XM_021071868.1 | XP_020927527.1 | 621 | 75 | |||
GenBank mRNA | AK234444.1 | - | 6 - 7 | 696 | 75 | |
AK238763.1 | - | 4 - 5 | 407 | 75 | ||
AK351475.1 | - | 4 - 5 | 398 | 75 | ||
AY609511.1 | - | 5 - 6 | 508 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |