Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190282.1 | NP_001177211.1 | 1 - 2 | 370 | 90 | |
GenBank mRNA | AK232886.1 | - | 180 | 90 | ||
AK236228.1 | - | 1 - 2 | 394 | 90 | ||
AY610153.1 | - | 1 - 2 | 446 | 90 | ||
HM046520.1 | - | 1 - 2 | 370 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |