Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193291.1 | NP_001180220.1 | 1 - 2 | 39 | 94 | |
GenBank mRNA | GU332640.1 | - | 1 - 2 | 39 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |