Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194976.1 | NP_001181905.1 | 5 - 6 | 656 | 62 | |
XM_005665209.3 | XP_005665266.1 | 768 | 62 | |||
XM_021093293.1 | XP_020948952.1 | 358 | 62 | |||
GenBank mRNA | AK344581.1 | - | 4 - 5 | 556 | 62 | |
GQ329714.1 | - | 5 - 6 | 656 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |