Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195324.1 | NP_001182253.1 | 2 - 3 | 2403 | 80 | |
XM_005655701.3 | XP_005655758.1 | 4297 | 80 | |||
XM_005655702.3 | XP_005655759.1 | 4301 | 80 | |||
XM_005655703.3 | XP_005655760.1 | 4270 | 80 | |||
XM_013988432.2 | XP_013843886.1 | 2720 | 80 | |||
XM_013988433.2 | XP_013843887.1 | 3109 | 80 | |||
XM_013988434.2 | XP_013843888.1 | 2483 | 80 | |||
XM_021090977.1 | XP_020946636.1 | 2606 | 80 | |||
XM_021090978.1 | XP_020946637.1 | 2579 | 80 | |||
XM_021090979.1 | XP_020946638.1 | 2616 | 80 | |||
XM_021090980.1 | XP_020946639.1 | 3095 | 80 | |||
XM_021090981.1 | XP_020946640.1 | 3099 | 80 | |||
XM_021090982.1 | XP_020946641.1 | 2510 | 80 | |||
XM_021090983.1 | XP_020946642.1 | 2520 | 80 | |||
GenBank mRNA | GU373701.1 | - | 2 - 3 | 2403 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |