Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195334.1 | NP_001182263.1 | 2 - 3 | 312 | 99 | |
GenBank mRNA | AK343347.1 | - | 2 - 3 | 357 | 99 | |
GU373715.1 | - | 2 - 3 | 312 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |