Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195345.1 | NP_001182274.1 | 6 - 7 | 778 | 83 | |
XM_021100301.1 | XP_020955960.1 | 1069 | 83 | |||
XM_021100302.1 | XP_020955961.1 | 815 | 83 | |||
GenBank mRNA | GU373703.1 | - | 6 - 7 | 778 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |