Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195351.1 | NP_001182280.1 | 2 - 3 | 213 | 95 | |
XM_005655892.3 | XP_005655949.1 | 1164 | 95 | |||
XM_005655894.3 | XP_005655951.1 | 636 | 95 | |||
GenBank mRNA | GU373672.1 | - | 2 - 3 | 213 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |