Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198923.2 | NP_001185852.2 | 4 - 5 | 765 | 98 | |
GenBank mRNA | AK393630.1 | - | 4 - 5 | 1017 | 98 | |
AK395655.1 | - | 4 - 5 | 737 | 98 | ||
AK400732.1 | - | 4 - 5 | 764 | 98 | ||
EU715290.1 | - | 4 - 5 | 694 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |