Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128492.1 | NP_001121964.1 | 3 - 4 | 375 | 86 | |
GenBank mRNA | AY610183.1 | - | 3 - 4 | 480 | 86 | |
EF397416.1 | - | 3 - 4 | 375 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |