Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244987.1 | NP_001231916.1 | 4 - 5 | 677 | 81 | |
XM_005655748.3 | XP_005655805.1 | 714 | 81 | |||
GenBank mRNA | AK234481.1 | - | 4 - 5 | 677 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |