Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245001.1 | NP_001231930.1 | 4 - 5 | 903 | 134 | |
XM_013997347.2 | XP_013852801.1 | 1149 | 131 | |||
XM_021088656.1 | XP_020944315.1 | 1959 | 131 | |||
XM_021088657.1 | XP_020944316.1 | 1958 | 131 | |||
GenBank mRNA | AK343398.1 | - | 4 - 5 | 903 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |