Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246205.1 | NP_001233134.1 | 1 - 2 | 442 | 144 | |
GenBank mRNA | AK233687.1 | - | 2 - 3 | 580 | 144 | |
AK391273.1 | - | 2 - 3 | 547 | 144 | ||
HQ266637.1 | - | 1 - 2 | 442 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |