Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206447.1 | NP_001193376.1 | 6 - 7 | 1909 | 94 | |
XM_013981389.2 | XP_013836843.2 | 2174 | 94 | |||
XM_021068276.1 | XP_020923935.1 | 2145 | 94 | |||
GenBank mRNA | AK351123.1 | - | 6 - 7 | 1907 | 94 | |
AK389431.1 | - | 6 - 7 | 1336 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |