Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206447.1 | NP_001193376.1 | 9 - 10 | 2286 | 69 | |
XM_021068276.1 | XP_020923935.1 | 2522 | 69 | |||
GenBank mRNA | AK351123.1 | - | 9 - 10 | 2284 | 69 | |
AK389431.1 | - | 9 - 10 | 1713 | 69 | ||
EW054906.2 | - | 1 - 2 | 91 | 69 | ||
EW509464.2 | - | 3 - 4 | 378 | 69 | ||
FS644374.1 | - | 2 - 3 | 245 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |