Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243255.1 | NP_001230184.1 | 3 - 4 | 567 | 69 | |
XM_021077227.1 | XP_020932886.1 | 520 | 69 | |||
GenBank mRNA | CV874307.1 | - | 3 - 4 | 371 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |