Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243360.1 | NP_001230289.1 | 4 - 5 | 352 | 68 | |
GenBank mRNA | AK350235.1 | - | 4 - 5 | 316 | 68 | |
BP144127.1 | - | 4 - 5 | 352 | 68 | ||
FD631484.1 | - | 1 - 2 | 152 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |