Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243410.1 | NP_001230339.1 | 2 - 3 | 338 | 119 | |
GenBank mRNA | AK389783.1 | - | 2 - 3 | 296 | 119 | |
AK392309.1 | - | 2 - 3 | 339 | 119 | ||
FS668818.1 | - | 2 - 3 | 339 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |