Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243455.1 | NP_001230384.1 | 1 - 2 | 283 | 91 | |
XM_005670934.3 | XP_005670991.1 | 294 | 91 | |||
GenBank mRNA | AK352109.1 | - | 1 - 2 | 134 | 91 | |
BP158014.1 | - | 1 - 2 | 283 | 91 | ||
CJ029038.1 | - | 1 - 2 | 138 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Sus scrofa Probe spans exons |