Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243455.1 | NP_001230384.1 | 2 - 3 | 424 | 111 | |
XM_005670934.3 | XP_005670991.1 | 435 | 111 | |||
XM_021071921.1 | XP_020927580.1 | 243 | 111 | |||
GenBank mRNA | AK352109.1 | - | 2 - 3 | 275 | 111 | |
BP158014.1 | - | 2 - 3 | 424 | 111 | ||
CJ029038.1 | - | 2 - 3 | 279 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Sus scrofa Probe spans exons |