Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243478.1 | NP_001230407.1 | 4 - 5 | 561 | 102 | |
GenBank mRNA | AK231488.1 | - | 4 - 5 | 563 | 102 | |
AK240399.1 | - | 4 - 5 | 480 | 102 | ||
AK346690.1 | - | 4 - 5 | 480 | 102 | ||
AY610201.1 | - | 4 - 5 | 537 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |