Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243488.1 | NP_001230417.1 | 4 - 5 | 541 | 59 | |
NM_001243489.1 | NP_001230418.1 | 4 - 5 | 534 | 59 | ||
XM_021079396.1 | XP_020935055.1 | 430 | 59 | |||
XM_021079397.1 | XP_020935056.1 | 607 | 59 | |||
GenBank mRNA | AK347170.1 | - | 4 - 5 | 534 | 59 | |
FS683329.1 | - | 4 - 5 | 541 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |