Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243595.1 | NP_001230524.1 | 6 - 7 | 876 | 79 | |
XM_021090951.1 | XP_020946610.1 | 1210 | 79 | |||
XM_021090952.1 | XP_020946611.1 | 821 | 79 | |||
XM_021090953.1 | XP_020946612.1 | 801 | 79 | |||
XM_021090954.1 | XP_020946613.1 | 825 | 79 | |||
GenBank mRNA | AK343638.1 | - | 6 - 7 | 876 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |