Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243622.1 | NP_001230551.1 | 5 - 6 | 720 | 89 | |
XM_021100397.1 | XP_020956056.1 | 868 | 89 | |||
XM_021100398.1 | XP_020956057.1 | 876 | 89 | |||
GenBank mRNA | AK232809.1 | - | 5 - 6 | 720 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |