Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM216B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243662.1 | NP_001230591.1 | 1 - 2 | 95 | 106 | |
XM_021065104.1 | XP_020920763.1 | 106 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |