Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243674.1 | NP_001230603.1 | 3 - 4 | 1863 | 70 | |
GenBank mRNA | AK344464.1 | - | 3 - 4 | 1877 | 70 | |
EW173110.2 | - | 2 - 3 | 321 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |