Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243716.1 | NP_001230645.1 | 2 - 3 | 1016 | 64 | |
GenBank mRNA | AK231710.1 | - | 2 - 3 | 1018 | 64 | |
AK350405.1 | - | 2 - 3 | 956 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |