Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RESF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243821.1 | NP_001230750.1 | 1 - 2 | 30 | 67 | |
XM_005664006.3 | XP_005664063.1 | 133 | 67 | |||
XM_005664007.3 | XP_005664064.1 | 133 | 67 | |||
XM_021090985.1 | XP_020946644.1 | 134 | 67 | |||
GenBank mRNA | AK344425.1 | - | 1 - 2 | 30 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |