Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243883.1 | NP_001230812.1 | 2 - 3 | 670 | 78 | |
GenBank mRNA | AJ236934.1 | - | 2 - 3 | 630 | 78 | |
AK231339.1 | - | 2 - 3 | 670 | 78 | ||
AK237000.1 | - | 2 - 3 | 636 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |