Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244093.1 | NP_001231022.1 | 3 - 4 | 278 | 108 | |
XM_013995256.2 | XP_013850710.1 | 809 | 108 | |||
GenBank mRNA | AK349993.1 | - | 3 - 4 | 269 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |