Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244259.1 | NP_001231188.1 | 3 - 4 | 355 | 53 | |
XM_021088566.1 | XP_020944225.1 | 594 | 53 | |||
XM_021088567.1 | XP_020944226.1 | 472 | 53 | |||
XM_021088568.1 | XP_020944227.1 | 341 | 53 | |||
GenBank mRNA | AK395673.1 | - | 4 - 5 | 409 | 53 | |
EU814891.1 | - | 3 - 4 | 355 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |