Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244416.1 | NP_001231345.1 | 2 - 3 | 527 | 69 | |
GenBank mRNA | AK347068.1 | - | 2 - 3 | 498 | 69 | |
AY610329.1 | - | 2 - 3 | 474 | 69 | ||
FS699491.1 | - | 2 - 3 | 456 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |