Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244447.1 | NP_001231376.1 | 1 - 2 | 180 | 64 | |
GenBank mRNA | AK231970.1 | - | 1 - 2 | 180 | 64 | |
AK233885.1 | - | 1 - 2 | 135 | 64 | ||
AK346764.1 | - | 1 - 2 | 135 | 64 | ||
AY609955.1 | - | 1 - 2 | 122 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |