Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244499.1 | NP_001231428.1 | 3 - 4 | 248 | 82 | |
GenBank mRNA | AK232668.1 | - | 3 - 4 | 229 | 82 | |
BP158234.1 | - | 3 - 4 | 204 | 82 | ||
BP997823.1 | - | 3 - 4 | 246 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |