Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ421947.1 | - | 753 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Triticum aestivum |