Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P5CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ005686.1 | - | 13 - 14 | 1590 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Vitis vinifera Probe spans exons |