Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY194361.1 | - | 2 - 3 | 329 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |