Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MADS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ504309.1 | - | 1 - 2 | 501 | 88 | |
GU133633.1 | - | 1 - 2 | 184 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Vitis vinifera Probe spans exons |