Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU591724.1 | - | 1 | 53 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Both primers and probe map within a single exon |