Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001205.1 | NP_001001205.1 | 5 - 6 | 522 | 79 | |
XM_018097691.1 | XP_017953180.1 | 4 - 5 | 1203 | 79 | ||
GenBank mRNA | BC067310.1 | - | 5 - 6 | 522 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |