Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sall4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001458.1 | NP_001001458.1 | 2 - 3 | 2494 | 118 | |
XM_012971614.2 | XP_012827068.1 | 2 - 3 | 2518 | 118 | ||
XM_012971616.2 | XP_012827070.1 | 2 - 3 | 3179 | 118 | ||
XM_018097690.1 | XP_017953179.1 | 2 - 3 | 2640 | 118 | ||
GenBank mRNA | BC067314.1 | - | 2 - 3 | 2494 | 118 | |
CT025472.2 | - | 2 - 3 | 2557 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |