Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sall4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001458.1 | NP_001001458.1 | 3 - 4 | 2788 | 71 | |
XM_012971614.2 | XP_012827068.1 | 3 - 4 | 2812 | 71 | ||
XM_012971615.2 | XP_012827069.1 | 3 - 4 | 2737 | 71 | ||
XM_012971616.2 | XP_012827070.1 | 3 - 4 | 3473 | 71 | ||
XM_018097690.1 | XP_017953179.1 | 3 - 4 | 2934 | 71 | ||
GenBank mRNA | BC067314.1 | - | 3 - 4 | 2788 | 71 | |
CT025472.2 | - | 3 - 4 | 2851 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |