Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aimp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004804.1 | NP_001004804.1 | 2 - 3 | 345 | 89 | |
XM_018097076.1 | XP_017952565.1 | 2 - 3 | 331 | 89 | ||
GenBank mRNA | BC074561.1 | - | 2 - 3 | 345 | 89 | |
CR760281.2 | - | 2 - 3 | 334 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |