Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmbim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004879.1 | NP_001004879.1 | 5 - 6 | 495 | 100 | |
GenBank mRNA | BC075267.1 | - | 5 - 6 | 495 | 100 | |
CR760482.2 | - | 5 - 6 | 457 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |