Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004898.1 | NP_001004898.1 | 1 - 2 | 124 | 103 | |
XM_012958820.2 | XP_012814274.1 | 2 - 3 | 277 | 103 | ||
XM_012958821.2 | XP_012814275.1 | 2 - 3 | 266 | 103 | ||
GenBank mRNA | BC075312.1 | - | 1 - 2 | 124 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |