Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004967.1 | NP_001004967.1 | 2 - 3 | 1202 | 63 | |
XM_012958832.2 | XP_012814286.1 | 2 - 3 | 1326 | 63 | ||
XM_018092029.1 | XP_017947518.1 | 2 - 3 | 1332 | 63 | ||
GenBank mRNA | BC075479.1 | - | 2 - 3 | 1202 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |